Learn More
Cayman Chemical NADPHsodIum salt 10mg
SDP
Catalog No. 502894914
Encompass_Preferred
The reduced form of the electron acceptor NADP+ and acts as an electron donor in various biological reactions
Catalog No. 50-289-4914 Supplier Cayman Chemical Supplier No. 900074310 MG
The reduced form of the electron acceptor NADP+ and acts as an electron donor in various biological reactions