Learn More
Cayman Chemical NADPHsodIum salt 50mg
SDP
Catalog No. 502892771
Encompass_Preferred
The reduced form of the electron acceptor NADP+ and acts as an electron donor in various biological reactions
Catalog No. 50-289-2771 Supplier Cayman Chemical Supplier No. 900074350 MG
The reduced form of the electron acceptor NADP+ and acts as an electron donor in various biological reactions