Learn More
Medchemexpress LLC NAE-IN-M22 10mg | 864420-54-2 | 363.32 g·mol-1 | C20H24Cl2N2 | 10 MG
SDP
Catalog No. 5000228364
Encompass_Preferred
NAE-IN-M22 is a potent, selective, and reversible inhibitor of the NEDD8-activating enzyme (NAE) used as a research tool to study neddylation-dependent signaling and apoptosis in cancer cell models. Supplied as a high-purity small-molecule solid, it is intended for laboratory research use only.
  • Inhibits NEDD8-activating enzyme in vitro and in vivo.
  • High purity (99.94%).
  • Supplied in mg-scale pack sizes, including 10 mg.
  • Suitable for studies of neddylation, ubiquitin-like modification, and apoptosis.
  • Store powder at -20°C for long-term stability; follow solvent storage recommendations for solutions.
Catalog No. 50-002-28364 Supplier Medchemexpress LLC Supplier No. HY11553710MG
NAE-IN-M22 is a potent, selective, and reversible inhibitor of the NEDD8-activating enzyme (NAE) used as a research tool to study neddylation-dependent signaling and apoptosis in cancer cell models. Supplied as a high-purity small-molecule solid, it is intended for laboratory research use only.
  • Inhibits NEDD8-activating enzyme in vitro and in vivo.
  • High purity (99.94%).
  • Supplied in mg-scale pack sizes, including 10 mg.
  • Suitable for studies of neddylation, ubiquitin-like modification, and apoptosis.
  • Store powder at -20°C for long-term stability; follow solvent storage recommendations for solutions.