Learn More
Medchemexpress LLC NAE-IN-M22 25mg | 864420-54-2 | 363.32 g/mol | C20H24Cl2N2 | 25 MG
SDP
Catalog No. 5000229100
Encompass_Preferred
A potent, selective, reversible inhibitor of the NEDD8 activating enzyme (NAE) with demonstrated antiproliferative and proapoptotic activity in cancer cell lines and reported in vivo tumor growth inhibition. Supplied as a white to off-white solid with high purity and high solubility in DMSO, it is available in multiple pack sizes and solution formats for research use.
  • Potent, selective, reversible NAE inhibition.
  • Demonstrated antiproliferative and proapoptotic activity in cells.
  • Effective in vivo tumor growth inhibition reported.
  • High purity solid with white to off-white appearance.
  • High solubility in DMSO and multiple stock concentration options.
Catalog No. 50-002-29100 Supplier Medchemexpress LLC Supplier No. HY11553725MG
A potent, selective, reversible inhibitor of the NEDD8 activating enzyme (NAE) with demonstrated antiproliferative and proapoptotic activity in cancer cell lines and reported in vivo tumor growth inhibition. Supplied as a white to off-white solid with high purity and high solubility in DMSO, it is available in multiple pack sizes and solution formats for research use.
  • Potent, selective, reversible NAE inhibition.
  • Demonstrated antiproliferative and proapoptotic activity in cells.
  • Effective in vivo tumor growth inhibition reported.
  • High purity solid with white to off-white appearance.
  • High solubility in DMSO and multiple stock concentration options.