Learn More
Medchemexpress LLC NAE-IN-M22 | 50MG
SDP
Catalog No. 5000227892
Encompass_Preferred
NAE-IN-M22 is a potent selective and reversible inhibitor of NEDD8 activating enzyme (NAE) with potency in micromolar range NAE-IN-M22 inhibits multiple cancer cell lines and induces apoptosis in A549 cells NAE-IN-M22 also can inhibit tumor growth in vivo[1]
Catalog No. 50-002-27892 Supplier Medchemexpress LLC Supplier No. HY11553750MG
NAE-IN-M22 is a potent selective and reversible inhibitor of NEDD8 activating enzyme (NAE) with potency in micromolar range NAE-IN-M22 inhibits multiple cancer cell lines and induces apoptosis in A549 cells NAE-IN-M22 also can inhibit tumor growth in vivo[1]