Learn More
TARGETMOL CHEMICALS INC NAE-IN-M22 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T88165MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NAE-IN-M22 (NEDD8 inhibitor M22) is a selective potent and reversible inhibitor of NEDD8 activating enzyme (NAE). NAE-IN-M22 inhibits multiple cancer cell lines and induces apoptosis in A549 cells. NAE-IN-M22 also can inhibit tumor growth in vivo. purity: 98%
Catalog No. 50-257-9044