Learn More
New England Biolabs, Inc. NaeI – 2500 units
SDP
Catalog No. 50812110
Encompass
A restriction endonuclease that recognizes the sequence GCC_^GGC.
Catalog No. 50-812-110 Supplier New England Biolabs, Inc. Supplier No. R0190L
A restriction endonuclease that recognizes the sequence GCC_^GGC.