Learn More
New England Biolabs, Inc. NaeI – 500 units
SDP
Catalog No. 50812111
Encompass
A restriction endonuclease that recognizes the sequence GCC_^GGC.
Catalog No. 50-812-111 Supplier New England Biolabs, Inc. Supplier No. R0190S
A restriction endonuclease that recognizes the sequence GCC_^GGC.