Learn More
Apexbio Technology LLC Nafamostat 81525-10-2 50mg
SDP
Catalog No. 5000092545
Encompass_Preferred
Nafamostat (CAS 81525-10-2) is a small-molecule inhibitor targeting a broad spectrum of serine proteases including thrombin plasmin kallikrein and human trypsin It is designed to block protease-mediated hydrolysis reactions thereby regulating coagulation pathways and inflammation-related protease activity Nafamostat exerts its biological activity primarily through reversible binding to protease active sites inhibiting their enzymatic functions In biochemical assays Nafamostat demonstrates potent inhibitory activity with an IC50 value of approximately 1 10 nM against human trypsin Based on these pharmacological properties Nafamostat holds research potential in coagulation pathway studies inflammation-related protease research pancreatitis modeling and mechanistic investigations of blood coagulation disorders and inflammatory responses
Catalog No. 50-000-92545 Supplier Apexbio Technology LLC Supplier No. B117750
Nafamostat (CAS 81525-10-2) is a small-molecule inhibitor targeting a broad spectrum of serine proteases including thrombin plasmin kallikrein and human trypsin It is designed to block protease-mediated hydrolysis reactions thereby regulating coagulation pathways and inflammation-related protease activity Nafamostat exerts its biological activity primarily through reversible binding to protease active sites inhibiting their enzymatic functions In biochemical assays Nafamostat demonstrates potent inhibitory activity with an IC50 value of approximately 1 10 nM against human trypsin Based on these pharmacological properties Nafamostat holds research potential in coagulation pathway studies inflammation-related protease research pancreatitis modeling and mechanistic investigations of blood coagulation disorders and inflammatory responses