Learn More
Medchemexpress LLC Nafamostat formate salt-13C6 | 98.1% | 1 MG
SDP
Catalog No. 5000405682
Encompass_Preferred
Nafamostat formate salt-13C6 is a 13C labeled Nafamostat, a synthetic serine protease inhibitor and anticoagulant. It suppresses T cell auto-reactivity by decreasing granzyme activity and CTL cytolysis, and also blocks activation of SARS-CoV-2. This compound is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Features a 13C labeled structure.
  • Acts as a synthetic serine protease inhibitor.
  • Functions as an anticoagulant.
  • Suppresses T cell auto-reactivity.
  • Blocks activation of SARS-CoV-2.
Catalog No. 50-004-05682 Supplier Medchemexpress LLC Supplier No. HYB0190S11MG
Add to Cart
Add to Cart
Nafamostat formate salt-13C6 is a 13C labeled Nafamostat, a synthetic serine protease inhibitor and anticoagulant. It suppresses T cell auto-reactivity by decreasing granzyme activity and CTL cytolysis, and also blocks activation of SARS-CoV-2. This compound is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Features a 13C labeled structure.
  • Acts as a synthetic serine protease inhibitor.
  • Functions as an anticoagulant.
  • Suppresses T cell auto-reactivity.
  • Blocks activation of SARS-CoV-2.