Learn More
Selleck Chemical LLC Nafamostat mesilate (FUT-175) S1386-10mM/1mL
SDP
Catalog No. 503111272
Encompass_Preferred
Nafamostat mesilate is a synthetic serine protease inhibitor used as an anticoagulant during hemodialysis Nafamostat mesylate blocks activation of SARS-CoV-2 and is investigated as a new treatment option for COVID-19 Nafamostat Mesilate attenuates inflammation and apoptosis
Catalog No. 50-311-1272 Supplier Selleck Chemical LLC Supplier No. S138610MM/1ML
Nafamostat mesilate is a synthetic serine protease inhibitor used as an anticoagulant during hemodialysis Nafamostat mesylate blocks activation of SARS-CoV-2 and is investigated as a new treatment option for COVID-19 Nafamostat Mesilate attenuates inflammation and apoptosis