Learn More
TARGETMOL CHEMICALS INC Nafamostat mesylate 100MG
SDP
Catalog No. 502877759
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Nafamostat mesylate (FUT-175), a synthetic serine protease inhibitor, has been used for the treatment of allergic disorders such as asthma, allergic rhinitis and atopic dermatitis. Purity 97.36%
Catalog No. 50-287-7759 Supplier TARGETMOL CHEMICALS INC Supplier No. T2392100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Nafamostat mesylate (FUT-175), a synthetic serine protease inhibitor, has been used for the treatment of allergic disorders such as asthma, allergic rhinitis and atopic dermatitis. Purity 97.36%