Learn More
Apexbio Technology LLC Nafamostat Mesylate(FUT-175), 10mg. Cas: 82956-11-4 MFCD: MFCD00941430. Serine protease inhibitor
SDP
Supplier:  Apexbio Technology LLC A258610
Encompass_Preferred
Nafamostat mesylate, previously known as FUT-175, is an inhibitor of serine protease that inhibits a variety of serine proteases, including trypsin and several proteases in the coagulation cascade. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4750