Learn More
Medchemexpress LLC Nafamostat mesylate | 82956-11-4 | >99.8% | 1 ML
SDP
Catalog No. 5000407383
Encompass_Preferred
Nafamostat mesylate (FUT-175) is an anticoagulant and a synthetic serine protease inhibitor. It demonstrates anticancer and antivirus effects. It induces apoptosis by up-regulating the expression of tumor necrosis factor receptor-1 (TNFR1) and can be utilized in the development of treatments for the pathological thickening of the arterial wall.
  • Anticoagulant
  • Synthetic serine protease inhibitor
  • Demonstrates anticancer effects
  • Demonstrates antivirus effects
  • Induces apoptosis by up-regulating the expression of tumor necrosis factor receptor-1 (TNFR1)
  • Can be utilized in the development of treatments for pathological thickening of the arterial wall
Catalog No. 50-004-07383 Supplier Medchemexpress LLC Supplier No. HYB0190A10MM/1ML
Add to Cart
Add to Cart
Nafamostat mesylate (FUT-175) is an anticoagulant and a synthetic serine protease inhibitor. It demonstrates anticancer and antivirus effects. It induces apoptosis by up-regulating the expression of tumor necrosis factor receptor-1 (TNFR1) and can be utilized in the development of treatments for the pathological thickening of the arterial wall.
  • Anticoagulant
  • Synthetic serine protease inhibitor
  • Demonstrates anticancer effects
  • Demonstrates antivirus effects
  • Induces apoptosis by up-regulating the expression of tumor necrosis factor receptor-1 (TNFR1)
  • Can be utilized in the development of treatments for pathological thickening of the arterial wall