Learn More
Medchemexpress LLC Nafamostat (mesylate) | 82956-11-4 | 99.85% | 25 MG
SDP
Catalog No. 5000451021
Encompass_Preferred
Nafamostat mesylate (FUT-175) is an anticoagulant and a synthetic serine protease inhibitor. It exhibits anticancer and antiviral effects, and induces apoptosis by up-regulating the expression of tumor necrosis factor-1 (TNFR1). It is also being investigated for treatments related to pathological thickening of the arterial wall.
  • Anticoagulant and synthetic serine protease inhibitor
  • Exhibits anticancer and antiviral effects
  • Induces apoptosis by up-regulating TNFR1 expression
  • Inhibits NF-κB activity
  • Suppresses invasiveness in Panc-1 cells
  • Demonstrates antiviral effects against Zika virus
  • Inhibits neointimal formation after carotid balloon injury
Catalog No. 50-004-51021 Supplier Medchemexpress LLC Supplier No. 25MGHYB0190A
Nafamostat mesylate (FUT-175) is an anticoagulant and a synthetic serine protease inhibitor. It exhibits anticancer and antiviral effects, and induces apoptosis by up-regulating the expression of tumor necrosis factor-1 (TNFR1). It is also being investigated for treatments related to pathological thickening of the arterial wall.
  • Anticoagulant and synthetic serine protease inhibitor
  • Exhibits anticancer and antiviral effects
  • Induces apoptosis by up-regulating TNFR1 expression
  • Inhibits NF-κB activity
  • Suppresses invasiveness in Panc-1 cells
  • Demonstrates antiviral effects against Zika virus
  • Inhibits neointimal formation after carotid balloon injury