Learn More
Medchemexpress LLC Nafamostat mesylate | 82956-11-4 | 99.9% | 500 MG
SDP
Catalog No. 5000405927
Encompass_Preferred
Nafamostat mesylate (FUT-175) is an anticoagulant and a synthetic serine protease inhibitor. It demonstrates anticancer and antiviral effects by inducing apoptosis through up-regulating the expression of tumor necrosis factor receptor-1 (TNFR1). This compound is also being investigated for its use in addressing the pathological thickening of the arterial wall.
  • Acts as an anticoagulant and synthetic serine protease inhibitor
  • Demonstrates anticancer and antiviral effects
  • Induces apoptosis via TNFR1 up-regulation
  • Investigated for treating pathological arterial wall thickening
  • Inhibits NF-κB activity and IκBα phosphorylation
  • Suppresses invasiveness in specific cell lines
  • Exhibits inhibitory effects on neointimal formation in animal models
  • Provides antiviral benefits against Zika virus in animal models
Catalog No. 50-004-05927 Supplier Medchemexpress LLC Supplier No. HYB0190A500MG
Nafamostat mesylate (FUT-175) is an anticoagulant and a synthetic serine protease inhibitor. It demonstrates anticancer and antiviral effects by inducing apoptosis through up-regulating the expression of tumor necrosis factor receptor-1 (TNFR1). This compound is also being investigated for its use in addressing the pathological thickening of the arterial wall.
  • Acts as an anticoagulant and synthetic serine protease inhibitor
  • Demonstrates anticancer and antiviral effects
  • Induces apoptosis via TNFR1 up-regulation
  • Investigated for treating pathological arterial wall thickening
  • Inhibits NF-κB activity and IκBα phosphorylation
  • Suppresses invasiveness in specific cell lines
  • Exhibits inhibitory effects on neointimal formation in animal models
  • Provides antiviral benefits against Zika virus in animal models