Learn More
Medchemexpress LLC Nafamostat mesylate (FUT-175) | 82956-11-4 | 99.9% | 100 MG
SDP
Catalog No. 5000406620
Encompass_Preferred
Nafamostat mesylate (FUT-175) is an anticoagulant and a synthetic serine protease inhibitor with demonstrated anticancer and antiviral effects. It induces apoptosis by up-regulating the expression of tumor necrosis factor receptor-1 (TNFR1) and is used in the development of treatments for pathological thickening of the arterial wall. This compound is intended for research use only.
  • Anticoagulant and synthetic serine protease inhibitor
  • Exhibits anticancer and antiviral effects
  • Induces apoptosis by up-regulating TNFR1 expression
  • Inhibits NF-κB activity
  • Suppresses invasiveness in Panc-1 cells
  • Demonstrates antiviral effects against Zika virus
  • Inhibits neointimal formation after balloon injury
Catalog No. 50-004-06620 Supplier Medchemexpress LLC Supplier No. HYB0190A100MG
Nafamostat mesylate (FUT-175) is an anticoagulant and a synthetic serine protease inhibitor with demonstrated anticancer and antiviral effects. It induces apoptosis by up-regulating the expression of tumor necrosis factor receptor-1 (TNFR1) and is used in the development of treatments for pathological thickening of the arterial wall. This compound is intended for research use only.
  • Anticoagulant and synthetic serine protease inhibitor
  • Exhibits anticancer and antiviral effects
  • Induces apoptosis by up-regulating TNFR1 expression
  • Inhibits NF-κB activity
  • Suppresses invasiveness in Panc-1 cells
  • Demonstrates antiviral effects against Zika virus
  • Inhibits neointimal formation after balloon injury