Learn More
Abcam Nafamostat Mesylate, Protease inhibitor, 50MG
Catalog No. 502706621
Encompass_Preferred
MW 539.6 Da, Purity >97%. Broad spectrum, potent protease inhibitor (Ki = 95.3 pM). Inhibits NF-κB activation and complement activation. Selective for human tryptase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6621 Supplier Abcam Supplier No. AB14143250MG
MW 539.6 Da, Purity >97%. Broad spectrum, potent protease inhibitor (Ki = 95.3 pM). Inhibits NF-κB activation and complement activation. Selective for human tryptase.
The product is subject to the following: Abcam Restricted Use Statement