Learn More
Apexbio Technology LLC Nafarelin Acetate 76932-56-4 50mg
SDP
Catalog No. 5000092688
Encompass_Preferred
Nafarelin Acetate is a synthetic agonist targeting gonadotropin-releasing hormone (GnRH) receptors It is designed to modulate the hypothalamic-pituitary-gonadal axis by initially stimulating and subsequently downregulating GnRH receptors in the pituitary gland thereby suppressing secretion of luteinizing hormone (LH) and follicle-stimulating hormone (FSH) Nafarelin Acetate exerts its biological activity primarily through receptor downregulation and suppression of pituitary hormone release Based on these pharmacological properties Nafarelin Acetate holds research potential in the investigation of endocrine signaling pathways associated with reproductive development disorders including central precocious puberty and endometriosis
Catalog No. 50-000-92688 Supplier Apexbio Technology LLC Supplier No. C644450
Nafarelin Acetate is a synthetic agonist targeting gonadotropin-releasing hormone (GnRH) receptors It is designed to modulate the hypothalamic-pituitary-gonadal axis by initially stimulating and subsequently downregulating GnRH receptors in the pituitary gland thereby suppressing secretion of luteinizing hormone (LH) and follicle-stimulating hormone (FSH) Nafarelin Acetate exerts its biological activity primarily through receptor downregulation and suppression of pituitary hormone release Based on these pharmacological properties Nafarelin Acetate holds research potential in the investigation of endocrine signaling pathways associated with reproductive development disorders including central precocious puberty and endometriosis