Learn More
Cayman Chemical NafcIlInsodIum salt hydrat 25g
SDP
Catalog No. 502982515
Encompass_Preferred
A semisynthetic and penicillinase-resistant penicillin antibiotic; active against 20 clinical isolates of S. aureus (MBCs = 0.078-0.312 UG/ml); reduces the number of kidney CFUs in a mouse model of S. aureus infection from 100-800 mg/kg; reduces the number of right atrial CFUs in a rabbit model of S. aureus-induced endocarditis
Catalog No. 50-298-2515 Supplier Cayman Chemical Supplier No. 2100825 G
A semisynthetic and penicillinase-resistant penicillin antibiotic; active against 20 clinical isolates of S. aureus (MBCs = 0.078-0.312 UG/ml); reduces the number of kidney CFUs in a mouse model of S. aureus infection from 100-800 mg/kg; reduces the number of right atrial CFUs in a rabbit model of S. aureus-induced endocarditis