Learn More
Medchemexpress LLC Nafcillin-d5 sodium | 1356354-25-0 | 97.8% | 1 MG
SDP
Catalog No. 5000408644
Encompass_Preferred
Nafcillin-d5 (sodium) is the deuterium labeled Nafcillin sodium. Nafcillin sodium, an antibiotic, is a reversible inhibitor of β-lactamase and can be used for the research of staphylococcal infections.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08644 Supplier Medchemexpress LLC Supplier No. HYB0555BS1MG
Nafcillin-d5 (sodium) is the deuterium labeled Nafcillin sodium. Nafcillin sodium, an antibiotic, is a reversible inhibitor of β-lactamase and can be used for the research of staphylococcal infections.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.