Learn More
Apexbio Technology LLC Nafcillin Sodium 7177-50-6 1g
SDP
Catalog No. 5000093205
Encompass_Preferred
Nafcillin Sodium (7177-50-6) is a small-molecule inhibitor targeting -lactamase It is designed to reversibly inhibit -lactamase thereby preventing bacterial resistance mediated by hydrolysis of -lactam antibiotics Nafcillin Sodium exerts its biological activity primarily through reversible inhibition of -lactamase In in vitro studies Nafcillin Sodium demonstrates inhibitory activity with an IC50 value of approximately 33 mM Based on these pharmacological properties Nafcillin Sodium holds research potential in investigating -lactamase-mediated antibiotic resistance mechanisms evaluating therapeutic combination strategies and studying bacterial susceptibility profiles
Catalog No. 50-000-93205 Supplier Apexbio Technology LLC Supplier No. B19831000
Nafcillin Sodium (7177-50-6) is a small-molecule inhibitor targeting -lactamase It is designed to reversibly inhibit -lactamase thereby preventing bacterial resistance mediated by hydrolysis of -lactam antibiotics Nafcillin Sodium exerts its biological activity primarily through reversible inhibition of -lactamase In in vitro studies Nafcillin Sodium demonstrates inhibitory activity with an IC50 value of approximately 33 mM Based on these pharmacological properties Nafcillin Sodium holds research potential in investigating -lactamase-mediated antibiotic resistance mechanisms evaluating therapeutic combination strategies and studying bacterial susceptibility profiles