Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nafcillin sodium monohydrate | 7177-50-6 | 98.1% | 100 MG
SDP

Catalog No. 5000408959
Encompass_Preferred

Nafcillin sodium monohydrate is an antibiotic and a reversible inhibitor of β-lactamase. It is used for the research of staphylococcal infections.

  • Targets: bacterial, antibiotic, beta-lactamase.
  • Pathway: anti-infection.
  • Solid storage: 4°C, sealed, away from moisture.
  • Solvent storage: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture).
  • In vitro solubility in H₂O: 5.83 mg/mL (12.83 mM), requires ultrasonic.
  • In vivo solubility in PBS: 100 mg/mL (220.04 mM), clear solution, requires ultrasonic.
  • In vivo solubility in 10% DMSO solutions: ≥ 2.5 mg/mL (5.50 mM), clear solution.
  • IC₅₀ target: β-lactam.
  • Exhibits bactericidal activity against methicillin-susceptible and methicillin-resistant *Staphylococcus aureus* in mice.

Catalog No. 50-004-08959 Supplier Medchemexpress LLC Supplier No. HYB0555A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nafcillin sodium monohydrate is an antibiotic and a reversible inhibitor of β-lactamase. It is used for the research of staphylococcal infections.

  • Targets: bacterial, antibiotic, beta-lactamase.
  • Pathway: anti-infection.
  • Solid storage: 4°C, sealed, away from moisture.
  • Solvent storage: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture).
  • In vitro solubility in H₂O: 5.83 mg/mL (12.83 mM), requires ultrasonic.
  • In vivo solubility in PBS: 100 mg/mL (220.04 mM), clear solution, requires ultrasonic.
  • In vivo solubility in 10% DMSO solutions: ≥ 2.5 mg/mL (5.50 mM), clear solution.
  • IC₅₀ target: β-lactam.
  • Exhibits bactericidal activity against methicillin-susceptible and methicillin-resistant *Staphylococcus aureus* in mice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.