Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nafcillin sodium monohydrate | 7177-50-6 | MFCD01941128 | 98.1% | 500 MG
SDP

Catalog No. 5000408535
Encompass_Preferred

Nafcillin sodium monohydrate is an antibiotic and a reversible inhibitor of β-lactamase, primarily used for the research of staphylococcal infections. For optimal stability, store this product at 4°C in a sealed container, away from moisture. When in solvent, it can be stored at -80°C for 6 months or -20°C for 1 month, also sealed and protected from moisture.

  • Functions as a reversible inhibitor of β-lactamase.
  • Used in research for staphylococcal infections.
  • Exhibits bactericidal activity against methicillin-susceptible Staphylococcus aureus (MSSA) and methicillin-resistant S. aureus (MRSA).
  • Molecular formula: C21H23N2NaO6S.
  • Molecular weight: 454.47.

Catalog No. 50-004-08535 Supplier Medchemexpress LLC Supplier No. HYB0555A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nafcillin sodium monohydrate is an antibiotic and a reversible inhibitor of β-lactamase, primarily used for the research of staphylococcal infections. For optimal stability, store this product at 4°C in a sealed container, away from moisture. When in solvent, it can be stored at -80°C for 6 months or -20°C for 1 month, also sealed and protected from moisture.

  • Functions as a reversible inhibitor of β-lactamase.
  • Used in research for staphylococcal infections.
  • Exhibits bactericidal activity against methicillin-susceptible Staphylococcus aureus (MSSA) and methicillin-resistant S. aureus (MRSA).
  • Molecular formula: C21H23N2NaO6S.
  • Molecular weight: 454.47.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.