Learn More
Medchemexpress LLC Nafcillin sodium monohydrate | 7177-50-6 | MFCD01941128 | 98.10% | 1 ML
SDP
Catalog No. 5000408428
Encompass_Preferred
Nafcillin sodium monohydrate is an antibiotic that functions as a reversible inhibitor of β-lactamase, making it suitable for research into staphylococcal infections.
  • Exhibits bactericidal activity against methicillin-susceptible Staphylococcus aureus (MSSA) and methicillin-resistant S. aureus (MRSA).
  • Has shown 100% protection in healthy mice with MSSA infections in an animal model.
  • Is currently being investigated in clinical trials for conditions such as osteomyelitis and staphylococcal bacteremia.
Catalog No. 50-004-08428 Supplier Medchemexpress LLC Supplier No. HYB0555A10MM/1ML
Nafcillin sodium monohydrate is an antibiotic that functions as a reversible inhibitor of β-lactamase, making it suitable for research into staphylococcal infections.
  • Exhibits bactericidal activity against methicillin-susceptible Staphylococcus aureus (MSSA) and methicillin-resistant S. aureus (MRSA).
  • Has shown 100% protection in healthy mice with MSSA infections in an animal model.
  • Is currently being investigated in clinical trials for conditions such as osteomyelitis and staphylococcal bacteremia.