Learn More
Thermo Scientific NAFION Tubing 0.3 mm ID
Supplier:  Thermo Scientific 0743390
Encompass
Sulfonated tetrafluorethylene copolymer tubing with 0.3mm inner diameter used as cation exchange membrane in analytical chemistry and electrochemistry applications provides high surface area to volume ratio.
Catalog No. NC9948632