Learn More
Selleck Chemical LLC Nafronyl oxalate salt S5472-25mg
SDP
Catalog No. 503108598
Encompass_Preferred
Nafronyl oxalate salt is the oxalate salt form of nafronyl which is a selective antagonist of 5-HT2 receptors and a vasodilator used in the management of peripheral and cerebral vascular disorders
Catalog No. 50-310-8598 Supplier Selleck Chemical LLC Supplier No. S547225MG
Nafronyl oxalate salt is the oxalate salt form of nafronyl which is a selective antagonist of 5-HT2 receptors and a vasodilator used in the management of peripheral and cerebral vascular disorders