Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nafronyl oxalate salt, MilliporeSigma™ Supelco™

Catalog No. 111013918 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3918 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3918 Supplier MilliporeSigma Supelco Supplier No. N13915G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 6-4-3200
Physical Form Neat
Linear Formula C24H33 NO3 · C2H2O4
MDL Number MFCD00079473
Synonym alpha-(1-Naphthylmethyl)-2-tetrahydrofuranpropionic acid diethylaminoethyl ester
Type Analytical Standard
Quantity 5 g
Formula Weight 473.56
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.