Learn More
TARGETMOL CHEMICALS INC Naftidrofuryl oxalate 500MG
SDP
Catalog No. 502878125
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Naftidrofuryl oxalate (Nafronyl oxalate) is a drug used in the management of peripheral and cerebral vascular disorders. It is claimed to enhance cellular oxidative capacity and to be a spasmolytic. It may also be an antagonist at 5HT-2 serotonin receptors. Purity 98.38%
Catalog No. 50-287-8125 Supplier TARGETMOL CHEMICALS INC Supplier No. T1081500MG
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Naftidrofuryl oxalate (Nafronyl oxalate) is a drug used in the management of peripheral and cerebral vascular disorders. It is claimed to enhance cellular oxidative capacity and to be a spasmolytic. It may also be an antagonist at 5HT-2 serotonin receptors. Purity 98.38%