Learn More
TARGETMOL CHEMICALS INC Naftifine hydrochloride 1G
SDP
Catalog No. 502876753
Encompass_Preferred
Also available in 1 mL, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Naftifine Hydrochloride is the hydrochloride salt form of naftifine, an allylamine derivate with synthetic broad-spectrum antifungal activity. Although the exact mechanism through which naftifine hydrochloride exerts its effect is unknown, it appears to selectively inhibit the enzyme squalene 2, 3-epoxidase, thereby inhibiting the biosynthesis of sterol. This results in a decreased amount of sterols, especially ergosterol which is the primary fungal membrane sterol, and a corresponding accumulation of squalene in fungal cells. Naftifine hydrochloride (Naftifine HCl) can be fungicidal as well as fungistatic to yeasts depending on the concentration and the organisms involved. Purity 99.76%
Catalog No. 50-287-6753 Supplier TARGETMOL CHEMICALS INC Supplier No. T15431G
Also available in 1 mL, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Naftifine Hydrochloride is the hydrochloride salt form of naftifine, an allylamine derivate with synthetic broad-spectrum antifungal activity. Although the exact mechanism through which naftifine hydrochloride exerts its effect is unknown, it appears to selectively inhibit the enzyme squalene 2, 3-epoxidase, thereby inhibiting the biosynthesis of sterol. This results in a decreased amount of sterols, especially ergosterol which is the primary fungal membrane sterol, and a corresponding accumulation of squalene in fungal cells. Naftifine hydrochloride (Naftifine HCl) can be fungicidal as well as fungistatic to yeasts depending on the concentration and the organisms involved. Purity 99.76%