Learn More
Medchemexpress LLC Naftifine hydrochloride | 65473-14-5 | 99.67% | 500 MG
SDP
Catalog No. 5000409334
Encompass_Preferred
Naftifine hydrochloride is an antibiotic with antifungal activity against dermatophytes, aspergilli, *Sporothrix schenckii*, and yeasts of the genus *Candida*. It can be used for research into superficial dermatomycoses inhibition, exhibiting activity against various fungal strains in vitro.
  • Antibiotic with antifungal activity
  • Effective against dermatophytes, aspergilli, *Sporothrix schenckii*, and *Candida* species
  • Suitable for research into superficial dermatomycoses inhibition
  • High purity of 99.67%
Catalog No. 50-004-09334 Supplier Medchemexpress LLC Supplier No. HYB0518A500MG
Naftifine hydrochloride is an antibiotic with antifungal activity against dermatophytes, aspergilli, *Sporothrix schenckii*, and yeasts of the genus *Candida*. It can be used for research into superficial dermatomycoses inhibition, exhibiting activity against various fungal strains in vitro.
  • Antibiotic with antifungal activity
  • Effective against dermatophytes, aspergilli, *Sporothrix schenckii*, and *Candida* species
  • Suitable for research into superficial dermatomycoses inhibition
  • High purity of 99.67%