Learn More
Medchemexpress LLC Naftifine (hydrochloride) | 65473-14-5 | 99.7% | 1 ML
SDP
Catalog No. 5000406868
Encompass_Preferred
Naftifine hydrochloride is an antibiotic with antifungal activity against dermatophytes, aspergilli, *Sporothrix schenckii*, and yeasts of the genus *Candida*. It is utilized for research into the inhibition of superficial dermatomycoses.
  • Antibiotic with antifungal activity
  • Effective against dermatophytes, aspergilli, *Sporothrix schenckii*, and *Candida* species
  • Utilized for research into inhibition of superficial dermatomycoses
  • Exhibits spectrum of activity with minimal inhibitory concentrations (MICs) against various fungi
  • For research use only
Catalog No. 50-004-06868 Supplier Medchemexpress LLC Supplier No. HYB0518A10MM/1ML
Naftifine hydrochloride is an antibiotic with antifungal activity against dermatophytes, aspergilli, *Sporothrix schenckii*, and yeasts of the genus *Candida*. It is utilized for research into the inhibition of superficial dermatomycoses.
  • Antibiotic with antifungal activity
  • Effective against dermatophytes, aspergilli, *Sporothrix schenckii*, and *Candida* species
  • Utilized for research into inhibition of superficial dermatomycoses
  • Exhibits spectrum of activity with minimal inhibitory concentrations (MICs) against various fungi
  • For research use only