Learn More
Medchemexpress LLC Naftifine hydrochloride | 65473-14-5 | 99.7% | 250 MG
SDP
Catalog No. 5000406807
Encompass_Preferred
Naftifine hydrochloride is an antibiotic with antifungal activity against dermatophytes, aspergilli, Sporothrix schenckii, and yeasts of the genus Candida. It can be used for the research of superficial dermatomycoses inhibition.
  • Antifungal activity against dermatophytes, aspergilli, and yeasts
  • Used for superficial dermatomycoses inhibition research
Catalog No. 50-004-06807 Supplier Medchemexpress LLC Supplier No. 250MGHYB0518A
Naftifine hydrochloride is an antibiotic with antifungal activity against dermatophytes, aspergilli, Sporothrix schenckii, and yeasts of the genus Candida. It can be used for the research of superficial dermatomycoses inhibition.
  • Antifungal activity against dermatophytes, aspergilli, and yeasts
  • Used for superficial dermatomycoses inhibition research