Learn More
Cayman Chemical Naftifine (hydrochloride), Purity: ≥98%, CAS: 65473-14-5, 100 mg
SDP
Catalog No. 502984091
Encompass_Preferred
Blocks ergosterol biosynthesis in fungi by inhibiting squalene epoxidase (Ki = 1.1 µM for C. albicans epoxidase)
Catalog No. 50-298-4091 Supplier Cayman Chemical Supplier No. 19234100 MG
Blocks ergosterol biosynthesis in fungi by inhibiting squalene epoxidase (Ki = 1.1 µM for C. albicans epoxidase)