Learn More
Medchemexpress LLC HY-B0391 500mg Medchemexpress, Naftopidil CAS:57149-07-2 Purity:>98%
SDP
Catalog No. 502082513
Encompass_Preferred
Medchemexpress, HY-B0391 500mg Naftopidil CAS:57149-07-2 Naftopidil (KT-611) is is a selective alpha1-adrenoceptor antagonist, with Kis of 3.7 nM, 20 nM and 1.2 nM for the cloned human α1a-, α1b- and α1d-adrenoceptor subtypes, respectively. Naftopidil has antiproliferative effects. Naftopidil can be used for the research of prostate hyperplasia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2513 Supplier Medchemexpress LLC Supplier No. HYB0391500MG
Medchemexpress, HY-B0391 500mg Naftopidil CAS:57149-07-2 Naftopidil (KT-611) is is a selective alpha1-adrenoceptor antagonist, with Kis of 3.7 nM, 20 nM and 1.2 nM for the cloned human α1a-, α1b- and α1d-adrenoceptor subtypes, respectively. Naftopidil has antiproliferative effects. Naftopidil can be used for the research of prostate hyperplasia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.