Learn More
Apexbio Technology LLC Naftopidil 57149-07-2 10g
SDP
Catalog No. 5000091513
Encompass_Preferred
Naftopidil (CAS 57149-07-2) is a small molecule that functions as a selective antagonist of 1-adrenergic receptors By blocking these receptors naftopidil inhibits adrenergic signaling associated with smooth muscle contraction in the lower urinary tract and vascular tissues In clinical studies administration of naftopidil has been associated with significant improvements in international prostatic symptom scores demonstrating its impact on lower urinary tract symptoms Naftopidil is primarily investigated for its antihypertensive properties and its potential to ameliorate symptoms associated with benign prostatic hyperplasia making it relevant for research on urological and cardiovascular disorders
Catalog No. 50-000-91513 Supplier Apexbio Technology LLC Supplier No. B136210000
Naftopidil (CAS 57149-07-2) is a small molecule that functions as a selective antagonist of 1-adrenergic receptors By blocking these receptors naftopidil inhibits adrenergic signaling associated with smooth muscle contraction in the lower urinary tract and vascular tissues In clinical studies administration of naftopidil has been associated with significant improvements in international prostatic symptom scores demonstrating its impact on lower urinary tract symptoms Naftopidil is primarily investigated for its antihypertensive properties and its potential to ameliorate symptoms associated with benign prostatic hyperplasia making it relevant for research on urological and cardiovascular disorders