Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Naftopidil | 57149-07-2 | 98.14% | 10 G
SDP

Catalog No. 5000406654
Encompass_Preferred

Naftopidil is a selective alpha1-adrenoceptor antagonist with Ki values of 3.7 nM, 20 nM, and 1.2 nM for the cloned human α1a-, α1b-, and α1d-adrenoceptor subtypes, respectively. It exhibits antiproliferative effects and is utilized in research concerning prostate hyperplasia.

  • Selective alpha1-adrenoceptor antagonist
  • Exhibits antiproliferative effects
  • Suitable for research related to prostate hyperplasia
  • Purity of 98.14%
  • Appears as a white to off-white solid
  • Soluble in DMSO; insoluble in water
  • Stable for 3 years at -20°C as powder
  • Stable for 2 years at -80°C in solvent

Catalog No. 50-004-06654 Supplier Medchemexpress LLC Supplier No. HYB039110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Naftopidil is a selective alpha1-adrenoceptor antagonist with Ki values of 3.7 nM, 20 nM, and 1.2 nM for the cloned human α1a-, α1b-, and α1d-adrenoceptor subtypes, respectively. It exhibits antiproliferative effects and is utilized in research concerning prostate hyperplasia.

  • Selective alpha1-adrenoceptor antagonist
  • Exhibits antiproliferative effects
  • Suitable for research related to prostate hyperplasia
  • Purity of 98.14%
  • Appears as a white to off-white solid
  • Soluble in DMSO; insoluble in water
  • Stable for 3 years at -20°C as powder
  • Stable for 2 years at -80°C in solvent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.