Learn More
Selleck Chemical LLC Naftopidil DiHCl S1387-25mg
SDP
Catalog No. 503111364
Encompass_Preferred
Naftopidil DiHCl (Flivas KT-611 Avishot BM-15275) is a selective 5-HT1A and 1-adrenergic receptor antagonist with IC50 of 0 1 M and 0 2 M respectively
Catalog No. 50-311-1364 Supplier Selleck Chemical LLC Supplier No. S138725MG
Naftopidil DiHCl (Flivas KT-611 Avishot BM-15275) is a selective 5-HT1A and 1-adrenergic receptor antagonist with IC50 of 0 1 M and 0 2 M respectively