Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naftopidil 98.0+%, TCI America™
SDP

Catalog No. N08325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08325G Supplier TCI America Supplier No. N08325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57149-07-2
Molecular Formula C24H28N2O3
Molecular Weight (g/mol) 392.50
MDL Number MFCD00242741,MFCD11046013
InChI Key HRRBJVNMSRJFHQ-UHFFFAOYNA-N
Synonym naftopidil, flivas, avishot, naftopidil inn, naftopidilum latin, naftopidil flivas, naftopidil unspecified, +--1-4-2-methoxyphenyl piperazinyl-3-1-naphthyloxy propan-2-ol, rs-1-4-2-methoxyphenyl-1-piperazinyl-3-1-naphthyloxy-2-propanol, 1-4-2-methoxyphenyl piperazin-1-yl-3-naphthalen-1-yloxy propan-2-ol
PubChem CID 4418
IUPAC Name 1-[4-(2-methoxyphenyl)piperazin-1-yl]-3-(naphthalen-1-yloxy)propan-2-ol
SMILES COC1=CC=CC=C1N1CCN(CC(O)COC2=C3C=CC=CC3=CC=C2)CC1

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 5 g
Formula Weight 392.50
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Naftopidil
RTECSNumber TL9336500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.