Learn More
Cayman Chemical NAG-thiazoline, Purity: ≥98%, CAS: 179030-22-9, 25 mg
SDP
Catalog No. 502948695
Encompass_Preferred
An OGA inhibitor (Ki = 180 nM); active against V. campbellii (MIC = 0.5 µM)
Catalog No. 50-294-8695 Supplier Cayman Chemical Supplier No. 3404425 MG
An OGA inhibitor (Ki = 180 nM); active against V. campbellii (MIC = 0.5 µM)