Learn More
Medchemexpress LLC NAG-thiazoline | 179030-22-9 | ≥95.0% | 219.26 | 100 MG
SDP
Catalog No. 5000349608
Encompass_Preferred
NAG-thiazoline is a potent O-GlcNAcase inhibitor with a Ki of 180 nM. It also demonstrates strong inhibition of GH20 GlcNAcase (VhGlcNAcase) with an IC₅₀ of 11.9 μM and a Ki of 62 μM. This compound effectively inhibits the growth of *Vibrio campbellii* strain 650 with a MIC value of 0.5 μM.
  • Targets bacterial O-GlcNAcase
  • Involved in anti-infection pathways
  • Plays a role in neuronal signaling pathways
Catalog No. 50-003-49608 Supplier Medchemexpress LLC Supplier No. HY139431100MG
Add to Cart
Add to Cart
NAG-thiazoline is a potent O-GlcNAcase inhibitor with a Ki of 180 nM. It also demonstrates strong inhibition of GH20 GlcNAcase (VhGlcNAcase) with an IC₅₀ of 11.9 μM and a Ki of 62 μM. This compound effectively inhibits the growth of *Vibrio campbellii* strain 650 with a MIC value of 0.5 μM.
  • Targets bacterial O-GlcNAcase
  • Involved in anti-infection pathways
  • Plays a role in neuronal signaling pathways