Learn More
Medchemexpress LLC NAG-thiazoline | 179030-22-9 | ≥95.0% | 219.26 | 25 MG
SDP
Catalog No. 5000349567
Encompass_Preferred
NAG-thiazoline is an O-GlcNAcase inhibitor with a Kᵢ of 180 nM. It is also a potent GH20 GlcNAcase (VhGlcNAcase) inhibitor with an IC₅₀ of 11.9 μM and a Kᵢ of 62 μM. It completely inhibits the growth of V. campbellii strain 650. This product is for research use only.
  • O-GlcNAcase inhibitor (Kᵢ of 180 nM)
  • Potent GH20 GlcNAcase (VhGlcNAcase) inhibitor (IC₅₀ of 11.9 μM, Kᵢ of 62 μM)
  • Inhibits the growth of V. campbellii strain 650 (MIC value of 0.5 μM)
  • Suitable for research applications
Catalog No. 50-003-49567 Supplier Medchemexpress LLC Supplier No. 25MGHY139431
NAG-thiazoline is an O-GlcNAcase inhibitor with a Kᵢ of 180 nM. It is also a potent GH20 GlcNAcase (VhGlcNAcase) inhibitor with an IC₅₀ of 11.9 μM and a Kᵢ of 62 μM. It completely inhibits the growth of V. campbellii strain 650. This product is for research use only.
  • O-GlcNAcase inhibitor (Kᵢ of 180 nM)
  • Potent GH20 GlcNAcase (VhGlcNAcase) inhibitor (IC₅₀ of 11.9 μM, Kᵢ of 62 μM)
  • Inhibits the growth of V. campbellii strain 650 (MIC value of 0.5 μM)
  • Suitable for research applications