Learn More
Medchemexpress LLC NAG-thiazoline | 179030-22-9 | ≥95.0% | 219.26 | 50 MG
SDP
Catalog No. 5000351154
Encompass_Preferred
NAG-thiazoline is an O-GlcNAcase inhibitor with a Ki of 180 nM. It is also a potent GH20 GlcNAcase (VhGlcNAcase) inhibitor with an IC50 of 11.9 μM and a Ki of 62 μM.
  • Completely inhibits the growth of V. campbellii strain 650 with a MIC value of 0.5 μM.
  • O-GlcNAcase inhibitor.
  • Potent GH20 GlcNAcase (VhGlcNAcase) inhibitor.
Catalog No. 50-003-51154 Supplier Medchemexpress LLC Supplier No. HY13943150MG
Add to Cart
Add to Cart
NAG-thiazoline is an O-GlcNAcase inhibitor with a Ki of 180 nM. It is also a potent GH20 GlcNAcase (VhGlcNAcase) inhibitor with an IC50 of 11.9 μM and a Ki of 62 μM.
  • Completely inhibits the growth of V. campbellii strain 650 with a MIC value of 0.5 μM.
  • O-GlcNAcase inhibitor.
  • Potent GH20 GlcNAcase (VhGlcNAcase) inhibitor.