Learn More
Medchemexpress LLC NAG-thiazoline | 179030-22-9 | MFCD22376563 | 98.1% | 219.26 g·mol-1 | C8H13NO4S | 5 MG
SDP
Catalog No. 5000223076
Encompass_Preferred
NAG-thiazoline is a small-molecule research reagent that inhibits O-GlcNAcase and shows activity against GH20 family GlcNAcases. It is supplied as a high-purity solid and is used as a biochemical tool for enzyme inhibition, assay development, and mechanistic studies. For research use only; consult the SDS for handling and storage.
Catalog No. 50-002-23076 Supplier Medchemexpress LLC Supplier No. HY1394315MG
NAG-thiazoline is a small-molecule research reagent that inhibits O-GlcNAcase and shows activity against GH20 family GlcNAcases. It is supplied as a high-purity solid and is used as a biochemical tool for enzyme inhibition, assay development, and mechanistic studies. For research use only; consult the SDS for handling and storage.