Learn More
TARGETMOL CHEMICALS INC NAG-thiazoline 25MG
SDP
Catalog No. 502878051
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. NAG-thiazoline (GlcNAc-thiazoline) is an O-GlcNAcase inhibitor with Ki value of 180 nM. It is active against V. campbellii with MIC value of 0.5 uM. Purity 99.35%
Catalog No. 50-287-8051 Supplier TARGETMOL CHEMICALS INC Supplier No. T919025MG
Add to Cart
Add to Cart
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. NAG-thiazoline (GlcNAc-thiazoline) is an O-GlcNAcase inhibitor with Ki value of 180 nM. It is active against V. campbellii with MIC value of 0.5 uM. Purity 99.35%