Learn More
Electron Microscopy Sciences Nageotte Chamber
SDP
Catalog No. 5027822
Encompass_Preferred
Bright line double ruling, 0.5 mm cell depth with coverglasses.
Catalog No. 50-278-22 Supplier Electron Microscopy Sciences Supplier No. 6351230
Bright line double ruling, 0.5 mm cell depth with coverglasses.