Learn More
Medchemexpress LLC HY-N3216 1mg Medchemexpress, Nagilactone B CAS:19891-51-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN32161MG
Encompass_Preferred
Medchemexpress, HY-N3216 1mg Nagilactone B CAS:19891-51-1 Nagilactone B, extracted from the root bark of Podocarpus nagi, is a liver X receptor ( LXR ) agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3937