Learn More
Cayman Chemical nagrelIdehydrochlorIde 10mg
SDP
Catalog No. 502994308
Encompass_Preferred
A PDE3 inhibitor (IC50 = 36 nM for the human platelet enzyme); inhibits thrombopoietin-induced megakaryocytopoiesis of isolated human CD34+ progenitor cells (IC50 = 26 nM); reduces ADP-, collagen-, thrombin-, or arachidonic acid-induced platelet aggregation in isolated rabbit platelet rich-plasma (EC50s = 0.31, 0.08, 0.18, and 0.1 UG/ml, respectively); inhibits platelet thrombus formation in a dog model of electrically induced carotid artery thrombosis when administered at 0.5-5 mg/kg
Catalog No. 50-299-4308 Supplier Cayman Chemical Supplier No. 2141110 MG
A PDE3 inhibitor (IC50 = 36 nM for the human platelet enzyme); inhibits thrombopoietin-induced megakaryocytopoiesis of isolated human CD34+ progenitor cells (IC50 = 26 nM); reduces ADP-, collagen-, thrombin-, or arachidonic acid-induced platelet aggregation in isolated rabbit platelet rich-plasma (EC50s = 0.31, 0.08, 0.18, and 0.1 UG/ml, respectively); inhibits platelet thrombus formation in a dog model of electrically induced carotid artery thrombosis when administered at 0.5-5 mg/kg