Learn More
AdipoGen NAI-112
SDP
Supplier:  AdipoGen AGCN20309M001
Encompass_Preferred
Chemical. CAS 1498289-64-7. Formula C109H147N25O31S2. MW 2367.6. Isolated from Actinoplanes sp. Labionin-containing class III lanthipeptide. Antinociceptive agent. Effective in animal models of nociceptic and inflammatory pain. Does not act through opioid, cannabinoid or descending adrenergic mechanisms. Most likely acts through the vanilloid-sensitive pathway by a novel target. It can be developed for the treatment of neuropathic pain. Weak antibacterial compound.
Catalog No. 50-256-2618