Learn More
AdipoGen NAI-802
SDP
Supplier:  AdipoGen AGCN20310M001
Encompass_Preferred
Chemical. CAS 1426209-02-0. Formula C90H141N25O26S4. MW 2117.5. Isolated from Actinoplanes sp. Actagardin-related class II lantibiotic. Antibacterial with improved activity than actagardine against Staphylococcus spp. and Streptococcus spp. Shows significant activity against Gram-positive anaerobic bacteria, such as Clost. difficile, Clostridium butyricum, Clostridium perfringens and Peptostreptococcus asaccharolyticus. Most probably inhibits cell wall synthesis and consequently bacterial growth by forming a complex with lipid intermediate II Lipid II, a key intermediate in peptidoglycan biosynthesis. Potential novel antimicrobial agent for the treatment of infections caused by Gram-positive pathogens including resistant strains.
Catalog No. 50-256-2616